accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3C(R)OH(S)OHNCH3H3C(E)

CAS RN 119567-63-4

IUPAC Name: (2S,3R,4E)-2-(dimethylamino)octadec-4-ene-1,3-diol
Synonyms: N,N-dimethylsphingosine N,N-dimethylsphing-4-enine
Molecular Weight (g/mol): 327.55
Molecular Formula: C20H41NO2
InChi Key: YRXOQXUDKDCXME-YIVRLKKSSA-N
SMILES: CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)N(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results