accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1187386-36-2

H3CNNBr

CAS RN 1187386-36-2

IUPAC Name: N-benzyl-5-bromo-N-methylpyridin-2-amine
Synonyms: N-benzyl-5-bromo-N-methylpyridin-2-amine
Molecular Weight (g/mol): 277.17
Molecular Formula: C13H13BrN2
InChi Key: WVDSURJOVZYTTF-UHFFFAOYSA-N
SMILES: CN(CC1=CC=CC=C1)C1=NC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results