CAS RN 1118460-77-7
CAS RN 1118460-77-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ UCPH 101
Selective non-substrate EAAT1 inhibitor
2
UCPH-101, MedChemExpress
MedChemExpress UCPH-101 is an excitatory amino acid transporter subtype 1 (EAAT1) inhibitor with an IC50 of 0.66 μM.