Learn More
Tocris Bioscience™ UCPH 101
Description
UCPH 101 is a selective non-substrate inhibitor of EAAT1 (IC50 values are 660, >300000 and >300000 nM for EAAT1, EAAT2 and EAAT3 respectively). Also demonstrates no significant inhibition at EAAT4 or EAAT5 in a patch-clamp electrophysiology assay (at final concentration up to 10 μM).
Specifications
Specifications
| Chemical Name or Material | 2-Amino-5,6,7,8-tetrahydro-4-(4-methoxyphenyl)-7-(naphthalen-1-yl)-5-oxo-4H-chromene-3-carbonitrile |
| CAS | 1118460-77-7 |
| Quantity | 10 mg |
| Target | Glutamate (EAAT) Transporter Inhibitors |
| Molecular Formula | C27H22N2O3 |
| Purity | 98% |