accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3H3CH3CCH3OHNNCH3H3CCH3CH3CH3H3COH

CAS RN 110546-20-8

IUPAC Name: 2,4-di-tert-butyl-6-({4-[(3,5-di-tert-butyl-2-hydroxyphenyl)methyl]piperazin-1-yl}methyl)phenol
Synonyms: 2,4-di-tert-butyl-6-({4-[(3,5-di-tert-butyl-2-hydroxyphenyl)methyl]piperazin-1-yl}methyl)phenol
Molecular Weight (g/mol): 522.82
Molecular Formula: C34H54N2O2
InChi Key: LAVRPTJDIGXPLJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(CN2CCN(CC3=C(O)C(=CC(=C3)C(C)(C)C)C(C)(C)C)CC2)=C(O)C(=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results