CAS RN 110374-16-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Desmethoxy Omeprazole, TRC
CAS: 110374-16-8 Molecular Formula: C16 H17 N3 O2 S Molecular Weight (g/mol): 315.39 Synonym: 2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole,Esomeprazole Magnesium Trihydrate Imp. B (EP),Omeprazole Imp. B (EP) IUPAC Name: 2-[(3,5-dimethylpyridin-2-yl)methylsulfinyl]-5-methoxy-1H-benzimidazole SMILES: COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)cc3C
2
2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.