accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1092460-71-3

IUPAC Name: 3-[3-methoxy-5-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: 3-[3-methoxy-5-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 246.19
Molecular Formula: C11H9F3O3
InChi Key: PHVYEXUYVALSAZ-UHFFFAOYSA-N
SMILES: COC1=CC(C=CC(O)=O)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results