accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NHOFFHCl

CAS RN 106266-04-0

IUPAC Name: 4-(2,4-difluorobenzoyl)piperidine hydrochloride
Synonyms: 4-(2,4-difluorobenzoyl)piperidine hydrochloride
Molecular Weight (g/mol): 261.7
Molecular Formula: C12H14ClF2NO
InChi Key: QPJONRGTWKXJLG-UHFFFAOYSA-N
SMILES: Cl.FC1=CC(F)=C(C=C1)C(=O)C1CCNCC1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results