accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 102940-86-3

FClOHO

CAS RN 102940-86-3

IUPAC Name: 2-chloro-3-fluorobenzoic acid
Synonyms: 2-chloro-3-fluorobenzoic acid
Molecular Weight (g/mol): 174.56
Molecular Formula: C7H4ClFO2
InChi Key: WJYAYXKXZNITAZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results