accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1024483-64-4

BrSOOHNFFF

CAS RN 1024483-64-4

IUPAC Name: 3-bromo-N-{[4-(trifluoromethyl)phenyl]methyl}benzene-1-sulfonamide
Synonyms: 3-bromo-N-{[4-(trifluoromethyl)phenyl]methyl}benzenesulfonamide
Molecular Weight (g/mol): 394.21
Molecular Formula: C14H11BrF3NO2S
InChi Key: RZKLDSNLHNWKFJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(CNS(=O)(=O)C2=CC(Br)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results