accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FHNF

CAS RN 1019613-18-3

IUPAC Name: 3-fluoro-N-[(4-fluorophenyl)methyl]aniline
Synonyms: 3-fluoro-N-[(4-fluorophenyl)methyl]aniline
Molecular Weight (g/mol): 219.24
Molecular Formula: C13H11F2N
InChi Key: HOWHCAHRSXJDGW-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2=CC(F)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results