accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFFFOCl

CAS RN 1017778-66-3

IUPAC Name: 2-methoxy-3-(trifluoromethyl)benzoyl chloride
Synonyms: 2-methoxy-3-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: VOQPCMLSRLWUEQ-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1C(F)(F)F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results