accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1017777-55-7

H3COClOHF

CAS RN 1017777-55-7

IUPAC Name: 3-chloro-5-fluoro-4-methoxyphenol
Synonyms: 3-chloro-5-fluoro-4-methoxyphenol
Molecular Weight (g/mol): 176.57
Molecular Formula: C7H6ClFO2
InChi Key: GHNZALBLIAFCPB-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(O)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results