accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CNClCH3

CAS RN 1010101-07-1

IUPAC Name: 2-chloro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-chloro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 253.53
Molecular Formula: C12H17BClNO2
InChi Key: FQLRMOOIOJBHPS-UHFFFAOYSA-N
SMILES: CC1=C(Cl)N=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results