Learn More
Invitrogen™ ZP1 Polyclonal Antibody

Description
Immunogen sequence: AGDQLIYENW LVSGIHIQKG PQGSITRDST FQLHVRCVFN ASDFLPIQAS IFPPPSPAPM TQPGPLRLEL RIAKDETFSS YYGEDDYPIV RLLREPVHVE VRLLQRTDPN LVLLLHQCWG APSANPFQQP QWPILSDGCP FKGDSYRTQM VALDGATPFQ SHYQRFTVAT FALLDSGSQR ALRGLVYLFC STSACHTSGL ETCSTACSTG TTRQRRSSGH RNDTARPQDI VSSPGPVGFE DSYGQEPTLG PTDSNGNSSL RPLLWAVLLL P; Positive Samples: OVCAR8, OVCAR-3, SKOV3, H460, A-549, Mouse lung, Rat ovary, Rat lung; Cellular Location: Cell membrane, Single-pass type I membrane protein, extracellular matrix, extracellular space.
Specifications
Specifications
| Antigen | ZP1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ZP1 |
| Gene Accession No. | O54766, P60852, Q62005 |
| Gene Alias | epididymis luminal protein 163; HEL163; mgc87693; OOMD; OOMD1; Processed zona pellucida sperm-binding protein 1; Zona pellucida glycoprotein 1; zona pellucida glycoprotein 1 (sperm receptor); zona pellucida sperm-binding protein 1; ZP1; Zp-1; zp1 protein |
| Gene Symbols | ZP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.