Learn More
Agilent Technologies ZORBAX SB-Phenyl, 3,0 x 100mm, 1,8 µm, 600 bar
Brand:  Agilent Technologies 828975-312
Encompass_Preferred
ZORBAX SB-Phenyl, 3,0 x 100mm, 1,8 µm, 600 bar
Product Code. 10554464
  • 1114.41 € / Each