missing translation for 'onlineSavingsMsg'
Learn More

Zoliflodacin, MedChemExpress

Product Code. 16447416
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447416

Brand: Medchem Express HY17647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zoliflodacin (ETX0914;AZD0914) is a novel spiropyrimidinetrione bacterial DNA gyrase/topoisomerase inhibitor. Zoliflodacin has potent in vitro antibacterial activity against Gram-positive and Gram-negative organisms, including S. aureus with the MIC90 of 0.25 μg/mL.

Zoliflodacin (ETX0914

Specifications

Chemical Name or Material Zoliflodacin
Physical Form Powder
CAS 1620458-09-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H22FN5O7
For Use With (Application) COVID-19-anti-virus
Synonym ETX0914 AZD0914
Solubility Information DMSO : 140 mg/mL (287.21 mM; Need ultrasonic)
SMILES O=C(NC(C12[C@]([C@H](C)O[C@H](C)C3)([H])N3C4=C(C=C5C(ON=C5N6C(OC[C@@H]6C)=O)=C4F)C2)=O)NC1=O
Molecular Weight (g/mol) 487.44
Formula Weight 487.44
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.