missing translation for 'onlineSavingsMsg'
Learn More

ZNF804A Antibody (ZNF804A/8945), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18590049 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18590049 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18590049 Supplier Novus Biologicals Supplier No. NBP324119AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF804A Monoclonal antibody specifically detects ZNF804A in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF804A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZNF804A/8945
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias C2orf10, ZNF804A zinc finger protein 804A
Host Species Mouse
Immunogen Recombinant full-length human ZNF804A protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91752
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.