missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF673 Polyclonal antibody specifically detects ZNF673 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNF673 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | FLJ20344zinc finger protein 673, protein ZNF673, putative zinc finger transcription factor, zinc finger family member 673 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human KRBOX4 (NP_060246.2). WQAAFIGKETLKDESGQESRTCRKSIYLSTEFDSVRQRLPKYYSWEKAFKTSFKLSWSKWKLCKKER |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?