missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF554 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541216
This item is not returnable.
View return policy
Description
Peptide sequence: GTPQASCSDW MTVLRNQDST YKKVALQEEP ASGINMIKLI REDGGWKQLE Sequence homology: Human: 100%.
ZNF554, or Zinc Finger Protein 554, is a member of the Kruppel-associated box domain-zinc finger proteins (KRAB-ZNF) subfamily, residing on chromosome 19p13. This region is characterized by a concentration of heterochromatin proteins, which indicates tightly regulated gene expression. ZNF554 is implicated in various cellular processes including transcription regulation and apoptosis. Dysregulation of ZNF554 has been linked to several diseases; notably, its decreased expression is associated with poor prognosis in gliomas, specifically grades III and IV astrocytomas, where it affects the PI3K-Akt signaling pathway. It also plays a role in placental development, with dysfunction contributing to conditions such as fetal growth restriction (FGR). Additionally, ZNF554 transcription is influenced by factors like ZEB1, ZEB2, and hypoxic conditions, with neighboring hypoxia-response elements showing hypermethylation in preeclampsia. This gene's expression can significantly alter the transcriptional landscape, with overexpression leading to the differential regulation of hundreds of genes, underscoring its importance in cellular regulation.
Specifications
| ZNF554 | |
| Polyclonal | |
| Unconjugated | |
| ZNF554 | |
| FLJ34817; Zinc finger protein 554; ZNF554 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 115196 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86TJ5 | |
| ZNF554 | |
| Synthetic peptide directed towards the N-terminal of human ZNF554 (aa 141-190). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction