Learn More
ZNF385D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16890385 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16890385 20 μL 20µL
16880385 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16890385 Supplier Proteintech Supplier No. 208061AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
ZNF385D gene ontology annotations related to this gene include nucleic acid binding; zinc ion binding.
Specifications
Antigen ZNF385D
Applications Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF385D
Gene Accession No. Q9H6B1
Gene Alias zinc finger protein 385D, Zinc finger protein 659, ZNF385D, ZNF659
Gene Symbols ZNF385D
Host Species Rabbit
Immunogen ZNF385D Fusion Protein Ag14746
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79750
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG