missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF366 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547653
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) ZNF24 and rhZNF206 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ZNF366 Polyclonal antibody specifically detects ZNF366 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
ZNF366 | |
Polyclonal | |
Unconjugated | |
Znf366 | |
DCSCRIPT; DC-SCRIPT; Dendritic cell-specific transcript protein; Zinc finger protein 366; ZNF366 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
167465 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8N895 | |
Znf366 | |
E. coli-derived recombinant human DC-SCRIPT/ZNF366 Met559-Ile744 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |