missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF34 Polyclonal Antibody

Description
Immunogen sequence: LSRPVPDQRPH KCDICEQSFE QRSYLNNHTR VHRS Highest antigen sequence identity to the following orthologs - mouse 46%, rat 46%.
May be involved in transcriptional regulation.
Specifications
Specifications
| Antigen | ZNF34 |
| Applications | ChIP Assay, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF34 |
| Gene Accession No. | Q8IZ26 |
| Gene Alias | KOX32; Zinc finger protein 34; zinc finger protein 34 (KOX 32); zinc finger protein KOX32; ZNF34 |
| Gene Symbols | ZNF34 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?