missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF300 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15907814
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15907814 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15907814 Supplier Invitrogen™ Supplier No. PA565037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QVTFVNSKTVT EASGHKYNPL GKIFQECIET DISIQRFHKY DAFKKNLKPN IDLPSCYKSN SRKKPDQSFG GGKSSSQSEP NSNLEKIHNG VIPFDDNQCG NVFRNTQSLI QYQNVETKEK SC Highest antigen sequence identity to the following orthologs - mouse 29%, rat 29%.

The protein encoded by this gene is a C2H2-type zinc finger DNA binding protein and likely transcriptional regulator. The function of this protein is not yet known. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF300
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF300
Gene Accession No. Q96RE9
Gene Alias kruppel-like zinc finger protein; zinc finger protein 300; ZNF300
Gene Symbols ZNF300
Host Species Rabbit
Immunogen Recombinant Human ZNF300. Recombinant protein control fragment (Product #RP-105803).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.