missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ZNF295 Antibody [PE], Novus Biologicals™

Product Code. 30034561 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30034561 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30034561 Supplier Novus Biologicals™ Supplier No. NB10074655PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF295 Polyclonal antibody specifically detects ZNF295 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF295
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias MGC15716, Zfp414, zinc finger protein 414
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human zinc finger protein 295 using the numbering given in entry NP_065778.3 (GeneID 49854).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49854
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.