Learn More
ZNF200 Monoclonal Antibody (OTI7C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808209
Description
Could have a role in spermatogenesis.Specifications
| ZNF200 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P98182 | |
| ZNF200 | |
| Full length human recombit protein of human ZNF200 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7C6 | |
| Unconjugated | |
| ZNF200 | |
| zinc finger protein 200 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7752 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.