missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF180 Monoclonal Antibody (4F3)

Description
Peptide Sequence: VAFTQRKAVI HERVCKSDET GEKSGLNSSL FSSPVIPIRN HFHKHVSHAK KWHLNAAVNS HQKINENETL YENNECGKPP QSIHLIQFTR TQTKDKSYGF SDRIQSFCHG.
Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See MIM 604749 for additional information on zinc finger proteins.
Specifications
Specifications
| Antigen | ZNF180 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4F3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF180 |
| Gene Accession No. | Q9UJW8 |
| Gene Alias | HHZ168; zinc finger protein 180; ZNF180 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?