missing translation for 'onlineSavingsMsg'
Learn More

ZM39923 hydrochloride, MedChemExpress

Product Code. p-200099987
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16431467

Brand: Medchem Express HY12589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZM39923 hydrochloride is a JAK3 inhibitor, with a pIC50 of 7.1; ZM39923 hydrochloride also potently inhibits tissue transglutaminase (TGM2) with an IC50 of 10 nM.

ZM39923 hydrochloride is a JAK3 inhibitor, with a pIC50 of 7.1

Specifications

Chemical Name or Material ZM39923 hydrochloride
Physical Form Solid
CAS 1021868-92-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H26ClNO
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 47 mg/mL (127.75 mM) ∣H2O : 1 mg/mL (2.72 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2C=CC=CC2=C1)CCN(C(C)C)CC3=CC=CC=C3.[H]Cl
Molecular Weight (g/mol) 367.91
Formula Weight 367.91
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.