missing translation for 'onlineSavingsMsg'
Learn More

ZM323881 hydrochloride, MedChemExpress

Product Code. 16496548
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496548

Brand: Medchem Express HY15467A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZM323881 hydrochloride is a potent and selective VEGFR2 inhibitor with an IC50 of less than 2 nM.

ZM323881 hydrochloride is a potent and selective VEGFR2 inhibitor with an IC50 of less than 2 nM.

Specifications

Chemical Name or Material ZM323881 hydrochloride
CAS 193000-39-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H19ClFN3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (121.40 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC1=C(O)C=C(NC2=C3C=CC(OCC4=CC=CC=C4)=CC3=NC=N2)C(F)=C1.Cl
Molecular Weight (g/mol) 411.86
Formula Weight 411.86
Percent Purity 98.6%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.