missing translation for 'onlineSavingsMsg'
Learn More

ZM 39923 hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18045992
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18045992 50 mg 50mg
18073912 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18045992 Supplier Tocris Bioscience Supplier No. 1367/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent, selective JAK3 inhibitor

Potent, selective inhibitor of Janus tyrosine kinase 3 (JAK3). pIC50 values are 7.1, 5.6, 4.4, and < 5.0 for JAK3, EGFR, JAK1 and CDK4 respectively. Breaks down in neutral buffer (t½ = 36 mins, pH 7.43, 25°C) to form ZM 449829 (Cat. No. 1366).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58753-54-1
Molecular Formula C23H26ClNO
Molecular Weight (g/mol) 367.917
InChI Key NJTUORMLOPXPBY-UHFFFAOYSA-N
PubChem CID 176406
IUPAC Name 3-[benzyl(propan-2-yl)amino]-1-naphthalen-2-ylpropan-1-one;hydrochloride
SMILES CC(C)N(CCC(=O)C1=CC2=CC=CC=C2C=C1)CC3=CC=CC=C3.Cl

Specifications

Quantity 50 mg
Synonym zm 39923 hydrochloride, jak3 inhibitor iv, zm39923 hydrochloride, 3-benzyl isopropyl amino-1-naphthalen-2-yl propan-1-one hydrochloride, 3-benzylisopropylamino-1-naphthalen-2-yl-propan-1-one hydrochloride, 2-naphthyl-n-isopropyl,n-benzyl-beta-aminoethylketone, hci, 3-n-benzyl-n-isopropyl amino-1-naphthalen-2-yl propan-1-one hydrochloride, c23h25no.clh
Formula Weight 367.92
Percent Purity >99%
Chemical Name or Material ZM 39923 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.