missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ ZLD 1039
GREENER_CHOICE

Product Code. p-7180144
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18786126

Brand: Tocris™ 5847/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective EZH2 inhibitor

ZLD 1039 is a potent and selective EZH2 inhibitor (IC50 = 5.6 nM). Displays selectivity for EZH2 over a panel of HMTs (inhibition less than 30% at 16 μM). Blocks H3K27 methylation in breast cancer cell lines. Induces tumor regression and metastasis in breast cancer xenograft-bearing mice. Reactivates silenced tumor suppressors in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[Ethyl(tetrahydro-2H-pyran-4-yl)amino]-N-[(2,3,5,6,7,8-hexahydro-1-methyl-3-oxo-4-isoquinolinyl)methyl]-2-methyl-5-[6-(4-methyl-1-piperazinyl)-3-pyridinyl]benzamide
CAS 1826865-46-6
Quantity 10 mg
Molecular Formula C36H48N6O3
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.