Learn More
WTW™ Zinc Test Kits
Product Code. p-8002906
Click to view available options
Type:
Zinc Extracting Reagent
Zinc Reagent Test Kit
Unit Size:
Each
Pack of 180
Product Code. 11747927
Brand: WTW™ 250452
Only null left
Add to basket
Add to basket
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For routine analysis in different applications
For Use With (Equipment) Photometers
No. of Tests 95
Sufficient For 25 Tests
Test Method DMP
Type Zinc Extracting Reagent