missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, Certified AR for Analysis, Fisher Chemical™

Product Code. 10726501 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10726501 500 g 500g
10294350 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10726501 Supplier Fisher Chemical Supplier No. Z/1350/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Color White
pH 7
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 81.38g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Zinc oxide
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.