missing translation for 'onlineSavingsMsg'
Learn More

Zinc formate, 98%

Product Code. p-7027452
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483257 100 g 100g
15493257 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483257 Supplier Thermo Scientific Alfa Aesar Supplier No. B24210.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-41-5 | C2H2O4Zn | 155.414 g/mol

Zinc formate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc formate is used as a pharmaceutical intermediate.

Solubility
Soluble in water (52 g/L at 20°C).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-41-5
Molecular Formula C2H2O4Zn
Molecular Weight (g/mol) 155.414
MDL Number MFCD00050836
InChI Key XOBMCBQSUCOAOC-UHFFFAOYSA-L
Synonym zinc formate, zinc diformate, formic acid, zinc salt, caswell no. 915, unii-zz67g2ct75, formic acid, zinc salt 2:1, epa pesticide chemical code 087802, 64-18-6 parent, bis-formyloxy zinc, zinc 2+ diformate
PubChem CID 11195
IUPAC Name zinc;diformate
SMILES C(=O)[O-].C(=O)[O-].[Zn+2]

Specifications

Quantity 100 g
UN Number UN3077
Solubility Information Soluble in water (52g/L at 20°C).
Formula Weight 155.4
Percent Purity 98%
Chemical Name or Material Zinc formate
compliance-icons compliance-icons
Product Identifier
  • Zinc formate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Zinc formate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.