missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
1 kg
6 x 250 g
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | ZnCl2 |
UN Number | 2331 |
Formula Weight | 136.3g/mol |
Percent Purity | ≥98% |
Grade | ACS/puriss. p.a. |
Vapor Pressure | 1mmHg (428°C) |
Chemical Name or Material | Zinc Chloride, Reag. Ph. Eur. reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'