missing translation for 'onlineSavingsMsg'
Learn More

Zimlovisertib, MedChemExpress

Product Code. 16473877
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473877

Brand: Medchem Express HY19836

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zimlovisertib (PF-06650833) is a potent, selective and orally active inhibitor of interleukin-1 receptor associated kinase 4 (IRAK4) with IC50s of 0.2 and 2.4 nM in the cell and PBMC assay, respectively. Zimlovisertib is used to treat diseases such as rheumatoid arthritis, lupus, and lymphomas.

Zimlovisertib (PF-06650833) is a potent, selective and orally active inhibitor of interleukin-1 receptor associated kinase 4 (IRAK4) with IC50s of 0.2 and 2.4 nM in the cell and PBMC assay, respectively. Zimlovisertib is used to treat diseases such as rheumatoid arthritis, lupus, and lymphomas.

Specifications

Chemical Name or Material Zimlovisertib
CAS 1817626-54-2
Color Light Nude
Purity Grade Notes Research
Molecular Formula C18H20FN3O4
Quantity 50 mg
Synonym PF-06650833
Solubility Information DMSO : 62.5 mg/mL (172.95 mM; ultrasonic and warming and heat to 60°C)
SMILES NC(C1=C(OC)C=C2C(C=CN=C2OC[C@@H]3[C@H](CC)[C@H](F)C(N3)=O)=C1)=O
Molecular Weight (g/mol) 361.37
Formula Weight 361.37
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.