missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika virus NS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-13196
This item is not returnable.
View return policy
Description
Zika virus NS3 Polyclonal antibody specifically detects Zika virus NS3 in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Zika virus NS3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin | |
| Flavivirin protease NS3 catalytic subunit, Non-structural protein 3, Serine protease NS3, ZIKV NS3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Zika virus NS3 (Zika virus (strain H/PF/2013)). The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction