missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zika Virus Envelope Protein Monoclonal Antibody (6931)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533302
This item is not returnable.
View return policy
Description
Does not cross react with the following viruses: Dengue 1-4, and Chikungunya virus.
As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. and Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
Specifications
| Zika Virus Envelope Protein | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 6931 | |
| Unconjugated | |
| Zika Envelope | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction