missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZIC5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17980043
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980043 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980043 Supplier Invitrogen™ Supplier No. PA5144306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZIC5 Polyclonal Antibody for IHC (P), ELISA

Zic5 (zinc finger protein of the cerebellum 5) is a C2H2 zinc finger transcription factor that influences development of the neural crest. Zic family members are abundant in developing and adult cerebellum. Zic family members are important during development, and have been associated with X-linked visceral heterotaxy and holoprosencephaly type 5. Zic5 is closely linked to Zic2, a related family member on chromosome 13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC5
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Zic5
Gene Accession No. Q96T25
Gene Alias 1700049L20Rik; Odd paired-related protein; odd-paired related; Opa-related protein; Opr; Zic family member 5; Zic family member 5 (odd-paired homolog, Drosophila); ZIC5; zinc family member 5 protein; zinc finger protein of the cerebellum 5; Zinc finger protein ZIC 5
Gene Symbols Zic5
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein ZIC 5 protein (217-276AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85416
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.