Learn More
Zibotentan, MedChemExpress
Product Code. 16455937
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16455937
Brand: Medchem Express HY10088
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Zibotentan (ZD4054) is a potent, selective and orally active endothelin A (ETA) receptor antagonist with a Ki of 13 nM. Zibotentan has no inhibitory effect on ETB. Zibotentan has anticancer effects and can be used for castration-resistant prostate cancer (CRPC) research.
Zibotentan (ZD4054) is a potent, selective and orally active endothelin A (ETA) receptor antagonist with a Ki of 13 nM. Zibotentan has no inhibitory effect on ETB. Zibotentan has anticancer effects and can be used for castration-resistant prostate cancer (CRPC) research.
Specifications
Chemical Name or Material Zibotentan
CAS 186497-07-4
Color White
Purity Grade Notes Research
Molecular Formula C19H16N6O4S
Quantity 5 mg
Synonym ZD4054
Solubility Information DMSO : 25 mg/mL (58.90 mM; Need ultrasonic)
SMILES O=S(NC1=NC=C(C)N=C1OC)(C2=CC=CN=C2C3=CC=C(C4=NN=CO4)C=C3)=O
Molecular Weight (g/mol) 424.43
Formula Weight 424.43
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death