missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZFYVE16 Polyclonal antibody specifically detects ZFYVE16 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Applications | Western Blot |
| Classification | Polyclonal |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
| Content And Storage | Store at 4°C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?