missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zfp72 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569739
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Zfp72 gene ontology annotations related to this gene include positive regulation by host of viral process; positive regulation by host of viral release from host cell; regulation of transcription, DNA-templated.
Specifications
| Zfp72 | |
| Polyclonal | |
| Unconjugated | |
| Zfp72 | |
| Zfp72; Zfp74; zinc finger protein 72; zinc finger protein 74 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 238722 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Zfp72 | |
| synthetic peptide directed towards the middle region of human Zfp72. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction