Learn More
Zeolite, MP Biomedicals™
Product Code. 11496301 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496301 500 g 500g
11486301 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11496301 Supplier MP Biomedicals Supplier No. 0219390290
Please to purchase this item. Need a web account? Register with us today!
Chemical Identifiers
CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O
Specifications
Physical Form Powder
Quantity 500 g
Density 2.1g/cm3 at 20°C
Chemical Name or Material Zeolite