Learn More
Thermo Scientific™ Zeiss 63X Objective, LD Plan-Neofluar (0.75 NA)
Product Code. 17256583 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17256583 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17256583 Supplier Thermo Scientific™ Supplier No. N010251
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
TRUSTED_SUSTAINABILITY
Specifications
Type Objective Lens
Product Line Zeiss
Description Zeiss 63X Objective, LD Plan-Neofluar (0.75 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 63X (0.75NA)
Quantity 1 each