missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Basement Membrane Marker Antibody (FIS 5F11/2), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50369AF594
This item is not returnable.
View return policy
Description
Zebrafish Basement Membrane Marker Monoclonal specifically detects Zebrafish Basement Membrane Marker in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Zebrafish Basement Membrane Marker | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Zebrafish Intestine | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| FIS 5F11/2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| Developmental Biology, Epitope Tags | |
| Zebrafish | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction