Learn More
Zeatin Riboside, TRC
Specifications
Specifications
| Chemical Name or Material | Zeatin Riboside |
| CAS | 28542-78-1 |
| Molecular Formula | C15 H21 N5 O5 |
| Quantity | 5 mg |
| Synonym | 6-(4-Hydroxy-3-methylbut-2-enylamino)-9-ß-D-ribofuranosylpurine, N6-(4-Hydroxy-3-methylbut-2-enyl)adenosine, 6-(4-Hydroxy-3-methyl-2-butenylamino)-9-β-D-ribofuranosylpurine, N-(4-Hydroxy-3-methyl-2-buten-1-yl)adenosine, N-(4-Hydroxy-3-methyl-2-butenyl)-adenosine |
| InChI Formula | InChI=1S/C15H21N5O5/c1-8(4-21)2-3-16-13-10-14(18-6-17-13)20(7-19-10)15-12(24)11(23)9(5-22)25-15/h2,6-7,9,11-12,15,21-24H,3-5H2,1H3,(H,16,17,18)/b8-2+/t9-,11+,12+,15-/m1/s1 |
| SMILES | C\C(=C/CNc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@H](O)[C@@H]3O)\CO |
| IUPAC Name | (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[[(E)-4-hydroxy-3-methylbut-2-enyl]amino]purin-9-yl]oxolane-3,4-diol |
| Molecular Weight (g/mol) | 351.358 |
| Formula Weight | 351.154 |