missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H12A Recombinant Rabbit Monoclonal Antibody (HL1442)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547045
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
ZC3H12A is an MCP1 -induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.
Specifications
| ZC3H12A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q5D1E8 | |
| ZC3H12A | |
| Recombinant protein encompassing a sequence within the center region of human ZC3H12A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Feline | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1442 | |
| Unconjugated | |
| ZC3H12A | |
| BC036563; bifunctional endoribonuclease and deubiquitinase ZC3H12A; dJ423B22.1; Endoribonuclease ZC3H12A; MCP-1 treatment-induced protein; MCP-induced protein 1; MCPIP; MCPIP1; Monocyte chemotactic protein-induced protein 1; Reg1; Regnase-1; RGD:1306776}; RGD1306776; ribonuclease ZC3H12A; RP3-423B22.1; zc3h12a; zc3h12a {ECO:0000312; Zinc finger CCCH domain-containing protein 12A; zinc finger CCCH type containing 12A; zinc finger CCCH-type containing 12A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 101088347, 80149 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction