Learn More
Invitrogen™ Human ZAP-70, His Tag Recombinant Protein
ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins.
Brand: Invitrogen™ P2782
Additional Details : Weight : 3.36000kg
Description
ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation.
Specifications
NP_001070 | |
7535 | |
20μg | |
SYK Kinase Family | |
Recombinant | |
Storage Buffer: 25 mM HEPES, pH 7.5, 150 mM NaCl, 0.5 mM DTT, 50% glycerol Store at -20°C. At first use, aliquot and store at -20°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
zeta-chain (TCR) associated protein kinase | |
Kinases & Inhibitors | |
Cytoplasmic Tyrosine Kinases |
Kinase Assay | |
70.1 kDa | |
ZAP-70 | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
ZAP70 | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Signal Transduction |