Learn More
Invitrogen™ Zagotenemab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542295
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Zagotenemab is targeting TAU protein, and it has launched a phase II clinical trial in April 2018.
Specifications
| Zagotenemab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LY-3303560 | |
| Human MAPT. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 0.61 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.