Learn More
Z433927330, MedChemExpress
Product Code. p-200099689
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16477977
Brand: Medchem Express HY126074
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Z433927330 is a potent and selective inhibitor of Aquaporin-7 (AQP7), less potently inhibits AQP3 and AQPs9, with IC50s of ∼0.2 μM, ∼0.7 μM and ∼1.1 μM for mAQP7, mAQP3 and mAQP9, respectively.
Z433927330 is a potent and selective inhibitor of Aquaporin-7 (AQP7), less potently inhibits AQP3 and AQPs9, with IC50s of ∼0.2 μM, ∼0.7 μM and ∼1.1 μM for mAQP7, mAQP3 and mAQP9, respectively.
Specifications
Chemical Name or Material Z433927330
Physical Form Solid
CAS 1005883-72-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H20N4O3
Solubility Information DMSO : ≥ 280 mg/mL (768.39 mM)
SMILES O=C(OCC)C1=CC=C(NC(NCC2=CC=C(N3N=CC=C3)C=C2)=O)C=C1
Molecular Weight (g/mol) 364.4
Formula Weight 364.4
Percent Purity 98.01%
Grade Research